This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ASTN2 SIRNA Assays ›
Gene Symbol:
ASTN2
Gene Name:
astrotactin 2
Gene Aliases:
bA67K19.1
Chromosome Location:
Chr.9: 116425225 - 117415039 on Build GRCh38
UniGene ID:
Hs.601562
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 137099

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ASTN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184734.1 NP_001171663.1 8 1319
NM_014010.4 NP_054729.3 22 3875
NM_198186.3 NP_937829.3 8 1640
NM_198187.3 NP_937830.3 8 1319
NM_198188.2 NP_937831.1 8 1319
GenBank mRNA AB014534.1 - 22 3875
AF116574.1 - 20 3096
AK024064.1 - 8 1290
AK127350.1 - 21 3096
AK315025.1 - 20 2939
BC018759.2 - 16 2324
BC029272.1 - 8 1640
BC093835.1 - 8 1153
BC101667.1 - 8 1152
BC143289.1 - 8 1153
BC143292.1 - 8 1153
BC143293.1 - 8 1153
BC146756.1 - 22 3875 9007192
BX649176.1 - 22 3210

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of protein localization to cell surface
calcium ion binding
inositol 1,3,4,5 tetrakisphosphate binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-47mq8:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline