This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other DOPEY2 SIRNA Assays ›
Gene Symbol:
DOPEY2
Gene Name:
dopey family member 2
Gene Aliases:
21orf5, C21orf5
Chromosome Location:
Chr.21: 36156781 - 36294274 on Build GRCh38
UniGene ID:
Hs.204575
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 137896

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: DOPEY2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320714.1 NP_001307643.1 8, 9 1195
NM_005128.3 NP_005119.2 8, 9 1086
XM_017028509.1 XP_016883998.1 5, 6 589
GenBank mRNA AB023150.2 - 5, 6 555
AJ237839.2 - 8, 9 1092

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

Golgi to endosome transport
endoplasmic reticulum organization
multicellular organism development
protein transport
cognition
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hldrw:80/100.66.76.65:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0