This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ATP6AP2 SIRNA Assays ›
Gene Symbol:
ATP6AP2
Gene Name:
ATPase H+ transporting accessory protein 2
Gene Aliases:
APT6M8-9, ATP6IP2, ATP6M8-9, ELDF10, HT028, M8-9, MRXE, MRXSH, MSTP009, PRR, RENR, XMRE, XPDS
Chromosome Location:
Chr.X: 40580964 - 40606637 on Build GRCh38
UniGene ID:
Hs.495960
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 138075

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ATP6AP2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005765.2 NP_005756.2 5 569
GenBank mRNA AF248966.1 - 5 574
AF291814.1 - 5 556
AK075382.1 - 5 627
AK296614.1 - 4 432
AK297760.1 - 4 543
AK314633.1 - 5 520
AK315948.1 - 4 404
AL049929.1 - 4 409
AY429341.1 - 4 432
BC010395.1 - 1 29
BC084541.1 - 5 549 6575732

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001007091, AB188298, BC079339.

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-zcg68:80/100.66.77.208:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline