siRNA Details
| Gene Symbol: SUCLA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003850.2 | NP_003841.1 | 2 | 217 | ||
| XM_017020806.1 | XP_016876295.1 | 2 | 437 | |||
| XM_017020807.1 | XP_016876296.1 | 2 | 179 | |||
| XM_017020808.1 | XP_016876297.1 | 2 | 204 | |||
| XM_017020809.1 | XP_016876298.1 | 2 | 204 | |||
| GenBank mRNA | AB035863.1 | - | 2 | 217 | ||
| AF058953.1 | - | 1 | 49 | |||
| AF087890.1 | - | 2 | 225 | |||
| AK001458.1 | - | 2 | 115 | |||
| AK001847.1 | - | 2 | 204 | |||
| AK225452.1 | - | 2 | 204 | |||
| AK299437.1 | - | 2 | 156 | |||
| AK302311.1 | - | 2 | 179 | |||
| AK315513.1 | - | 2 | 179 | |||
| AK316041.1 | - | 2 | 184 | |||
| AK316271.1 | - | 3 | 532 | |||
| BC027587.2 | - | 2 | 169 | 4829291 | ||
| BX640602.1 | - | 2 | 188 | |||
| KU178442.1 | - | 2 | 160 | |||

SUCLA2
Genomic Map