This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TYRO3 SIRNA Assays ›
Gene Symbol:
TYRO3
Gene Name:
TYRO3 protein tyrosine kinase
Gene Aliases:
BYK, Dtk, Etk-2, RSE, Rek, Sky, Tif
Chromosome Location:
Chr.15: 41559022 - 41579338 on Build GRCh38
UniGene ID:
Hs.381282
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 139605

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TYRO3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006293.3 NP_006284.2 9 1408
XM_017022542.1 XP_016878031.1 9 1373
XM_017022543.1 XP_016878032.1 9 1209
GenBank mRNA AB209431.1 - 9 1437
AK022124.1 - 1 2074
BC029925.2 - 1 58
BC034960.1 - 9 1190
BC049368.1 - 9 1182 4821587
BC051756.1 - 9 1187 6095596
D17517.1 - 9 1396
D50479.1 - 9 1168
U02566.1 - 6 619
U05682.1 - 9 1178

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

natural killer cell differentiation
cell adhesion
signal transduction
spermatogenesis
phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
forebrain cell migration
signal transduction by protein phosphorylation
platelet activation
secretion by cell
negative regulation of toll-like receptor signaling pathway
substrate adhesion-dependent cell spreading
ovulation cycle
apoptotic cell clearance
protein kinase B signaling
negative regulation of neuron apoptotic process
negative regulation of innate immune response
viral entry into host cell
protein autophosphorylation
negative regulation of inflammatory response
negative regulation of lymphocyte activation
vagina development
neuron cellular homeostasis
platelet aggregation
virus receptor activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
protein binding
ATP binding
phosphatidylinositol 3-kinase binding
protein heterodimerization activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0