This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other RMDN3 SIRNA Assays ›
Gene Symbol:
RMDN3
Gene Name:
regulator of microtubule dynamics 3
Gene Aliases:
FAM82A2, FAM82C, RMD-3, RMD3, ptpip51
Chromosome Location:
Chr.15: 40735884 - 40755336 on Build GRCh38
UniGene ID:
Hs.511067
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 140511

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: RMDN3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304802.1 NP_001291731.1 13 1813
NM_001323894.1 NP_001310823.1 13 1727
NM_001323895.1 NP_001310824.1 13 1944
NM_001323896.1 NP_001310825.1 13 1805
NM_001323897.1 NP_001310826.1 13 1921
NM_018145.2 NP_060615.1 13 1843
XM_011521755.2 XP_011520057.1 10 2089
GenBank mRNA AK001441.1 - 13 1767
AK025963.1 - 10 1156
AK090731.1 - 6 1715
AK092058.1 - 13 1726
AK097286.1 - 14 1655
AK123192.1 - 14 1720
AK123282.1 - 12 2650
AY358793.1 - 12 1583
BC008970.2 - 9 1040
BC063844.1 - 13 1731 6527521

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cellular calcium ion homeostasis
apoptotic process
cell differentiation
protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7988999796-smrv7:80/100.66.72.156:80.
git-commit: a642765d7314f3190119cdf516b7ea8f1d23272c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.1-2025.11.99-1.0