This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other VPS25 SIRNA Assays ›
Gene Symbol:
VPS25
Gene Name:
vacuolar protein sorting 25 homolog
Gene Aliases:
DERP9, EAP20, FAP20
Chromosome Location:
Chr.17: 42773436 - 42779600 on Build GRCh38
UniGene ID:
Hs.500165
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141094

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: VPS25
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032353.3 NP_115729.1 6 622
GenBank mRNA BC006282.2 - 6 622 4025884

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001173451, FQ220187, FQ229682, FQ233075, FQ213477.
This siRNA also targets the following transcripts in Mouse: NM_026776, NM_001284412, NM_001284411, AK217046, AK002500, and others.

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
autophagy
endosomal transport
multivesicular body assembly
protein targeting to vacuole involved in ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
regulation of viral life cycle
structural molecule activity
protein binding
protein homodimerization activity
protein N-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline