This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other INF2 SIRNA Assays ›
Gene Symbol:
INF2
Gene Name:
inverted formin, FH2 and WH2 domain containing
Gene Aliases:
C14orf151, C14orf173, CMTDIE, FSGS5, pp9484
Chromosome Location:
Chr.14: 104689606 - 104719610 on Build GRCh38
UniGene ID:
Hs.24956
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141127

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: INF2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032714.2 NP_116103.1 5 1240
GenBank mRNA BC006173.1 - 5 1225

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

actin cytoskeleton organization
regulation of mitochondrial fission
actin binding
Rho GTPase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-55586bc97b-stzfq:80/100.66.77.128:80.
git-commit: d8c775737291b1e1a3c617895b4ca460ad723db2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0