This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other COQ10A SIRNA Assays ›
Gene Symbol:
COQ10A
Gene Name:
coenzyme Q10A
Gene Aliases:
-
Chromosome Location:
Chr.12: 56266858 - 56270966 on Build GRCh38
UniGene ID:
Hs.4096
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141301

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: COQ10A
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099337.1 NP_001092807.1 2 267
NM_144576.3 NP_653177.3 2 511
GenBank mRNA AK057003.1 - 2 427
AK057014.1 - 1 228
AY358861.1 - 2 199
BC002435.2 - 1 88
BC026922.1 - 2 267
BC047444.1 - 2 239
BC073923.1 - 2 395 5188138
DB271789.1 - 2 511

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108727, XM_006240759.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-wtj7v:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline