siRNA Details
| Gene Symbol: FSIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001324338.1 | NP_001311267.1 | 10 | 1251 | ||
| NM_152597.4 | NP_689810.3 | 10 | 1348 | |||
| XM_011521305.2 | XP_011519607.1 | 10 | 1224 | |||
| XM_011521306.1 | XP_011519608.1 | 10 | 1637 | |||
| XM_011521307.2 | XP_011519609.1 | 10 | 1224 | |||
| XM_011521309.2 | XP_011519611.1 | 10 | 1224 | |||
| XM_011521311.2 | XP_011519613.1 | 6 | 1908 | |||
| XM_017021972.1 | XP_016877461.1 | 6 | 701 | |||
| GenBank mRNA | AK093308.1 | - | 10 | 1330 | ||
| AY260140.1 | - | 9 | 1145 | |||
| BC045191.2 | - | 10 | 1340 | 5297527 | ||

FSIP1
Genomic Map