This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other ANKFN1 SIRNA Assays ›
Gene Symbol:
ANKFN1
Gene Name:
ankyrin repeat and fibronectin type III domain containing 1
Gene Aliases:
-
Chromosome Location:
Chr.17: 56110935 - 56517016 on Build GRCh38
UniGene ID:
Hs.673040
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141589

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: ANKFN1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153228.2 NP_694960.2 8, 9 1049
XM_006721728.3 XP_006721791.1 7, 8 1272
XM_011524428.2 XP_011522730.1 8, 9 1049
XM_011524429.2 XP_011522731.1 8, 9 1206
XM_011524430.2 XP_011522732.1 9, 10 1172
XM_011524431.2 XP_011522733.1 8, 9 1104
XM_011524433.2 XP_011522735.1 2, 3 462
XM_017024263.1 XP_016879752.1 10, 11 1266
XM_017024264.1 XP_016879753.1 10, 11 2251
XM_017024265.1 XP_016879754.1 10, 11 1460
XM_017024266.1 XP_016879755.1 8, 9 1221
XM_017024267.1 XP_016879756.1 7, 8 1073
XM_017024268.1 XP_016879757.1 9, 10 2157
XM_017024269.1 XP_016879758.1 7, 8 1346
XM_017024270.1 XP_016879759.1 10, 11 1264
GenBank mRNA AK095654.1 - 8, 9 1049

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0