This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other CHN2 SIRNA Assays ›
Gene Symbol:
CHN2
Gene Name:
chimerin 2
Gene Aliases:
ARHGAP3, BCH, CHN2-3, RHOGAP3
Chromosome Location:
Chr.7: 29146547 - 29514335 on Build GRCh38
UniGene ID:
Hs.654611
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 14167

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: CHN2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001293069.1 NP_001279998.1 6, 7 560
NM_001293070.1 NP_001279999.1 6, 7 774
NM_001293071.1 NP_001280000.1 4, 5 679
NM_001293072.1 NP_001280001.1 5, 6 492
NM_004067.3 NP_004058.1 5, 6 735
XM_011515105.2 XP_011513407.1 8, 9 615
XM_011515106.2 XP_011513408.1 7, 8 576
XM_011515107.2 XP_011513409.1 7, 8 1358
XM_017011721.1 XP_016867210.1 9, 10 633
XM_017011722.1 XP_016867211.1 8, 9 1376
GenBank mRNA AK124021.1 - 1, 2 1843
AK293959.1 - 6, 7 354
AK294032.1 - 5, 6 492
AK294271.1 - 6, 7 561
AK313021.1 - 5, 6 509
BC112155.1 - 5, 6 480 8327623
DA190643.1 - 6, 7 560
EU732760.1 - 4, 5 242
EU732765.1 - 5, 6 298
EU732766.1 - 5, 6 298
GQ924106.1 - 6, 7 561
U28926.1 - 5, 6 532

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

positive regulation of signal transduction
intracellular signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
SH3/SH2 adaptor activity
GTPase activator activity
protein binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline