This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GPR180 SIRNA Assays ›
Gene Symbol:
GPR180
Gene Name:
G protein-coupled receptor 180
Gene Aliases:
ITR
Chromosome Location:
Chr.13: 94601850 - 94634645 on Build GRCh38
UniGene ID:
Hs.439363
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141850

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GPR180
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_180989.5 NP_851320.1 3 394
GenBank mRNA AK126794.1 - 3 299
AK289850.1 - 3 641
BC033906.1 - 3 369
BC052243.1 - 3 341 6083073
CD110068.1 - 3 421

Back To Top

More Information


Gene Ontology Categories:

Function(s)

G-protein coupled receptor signaling pathway
response to pheromone

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-5d9f4b9bfc-tdbx5:80/100.66.78.52:80.
git-commit: 52aea788d112809596cd23e639b0bc7998ca5a01
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.40.1-2025.11.48-1.0