This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TMEM119 SIRNA Assays ›
Gene Symbol:
TMEM119
Gene Name:
transmembrane protein 119
Gene Aliases:
OBIF
Chromosome Location:
Chr.12: 108589846 - 108598118 on Build GRCh38
UniGene ID:
Hs.449718
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 141866

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 274.65
274.65
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TMEM119
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181724.2 NP_859075.2 2 1310
XM_011538271.2 XP_011536573.1 3 1500
GenBank mRNA AK075501.1 - 2 1200
AK097290.1 - 4 805
AK301202.1 - 3 964
AY358389.1 - 2 1309
BC096825.1 - 2 1221 30558500
ES311302.1 - 1 21

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
spermatogenesis
biological_process
positive regulation of bone mineralization
biomineral tissue development
positive regulation of osteoblast proliferation
positive regulation of osteoblast differentiation
spermatid differentiation
positive regulation of bone development
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-2mxx6:80/100.66.73.198:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0