siRNA Details
| Gene Symbol: MNDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002432.1 | NP_002423.1 | 5 | 800 | ||
| GenBank mRNA | AK290392.1 | - | 5 | 807 | ||
| AK313210.1 | - | 5 | 811 | |||
| BC020715.1 | - | 5 | 838 | |||
| BC032319.1 | - | 5 | 811 | 5223430 | ||
| M81750.1 | - | 5 | 800 | |||

MNDA
Genomic Map