This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NEK3 SIRNA Assays ›
Gene Symbol:
NEK3
Gene Name:
NIMA related kinase 3
Gene Aliases:
HSPK36
Chromosome Location:
Chr.13: 52132643 - 52159861 on Build GRCh38
UniGene ID:
Hs.409989
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 143615

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NEK3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146099.1 NP_001139571.1 2 458
NM_002498.2 NP_002489.1 2 458
NM_152720.2 NP_689933.1 2 477
NR_027415.2 2 458
GenBank mRNA AB072828.1 - 1 127
AI209040.1 - 2 320
AK131359.1 - 1 1376
AK290259.1 - 2 190
AL833951.1 - 2 317
BC019916.1 - 2 185 5000918
BQ648157.1 - 2 394
DA733733.1 - 2 458
DB185803.1 - 2 455

Back To Top

More Information


Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
microtubule-based process
chromosome segregation
mitotic nuclear division
establishment of cell polarity
neuron projection morphogenesis
cell division
regulation of tubulin deacetylation
protein serine/threonine kinase activity
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-zvvnw:80/100.66.74.101:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline