siRNA Details
| Gene Symbol: MUC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145006.1 | NP_001138478.1 | 4 | 370 | ||
| NM_001145007.1 | NP_001138479.1 | 4 | 309 | |||
| NM_152291.2 | NP_689504.2 | 3 | 272 | |||
| GenBank mRNA | AK093828.1 | - | 3 | 192 | ||
| AK312855.1 | - | 3 | 191 | |||
| BC025688.1 | - | 3 | 170 | 5205526 | ||
| BX647190.1 | - | 3 | 191 | |||
| DB194707.1 | - | 4 | 370 | |||
| DB199851.1 | - | 4 | 309 | |||
| DB206328.1 | - | 3 | 272 | |||
| L13283.1 | - | 3 | 178 | |||

MUC7
Genomic Map