This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
See other EPB41 SIRNA Assays ›
Gene Symbol:
EPB41
Gene Name:
erythrocyte membrane protein band 4.1
Gene Aliases:
4.1R, EL1, HE
Chromosome Location:
Chr.1: 28887091 - 29120046 on Build GRCh38
UniGene ID:
Hs.175437
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146879
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EPB41
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001166005.1
NP_001159477.1
2
362
NM_001166006.1
NP_001159478.1
2
435
NM_001166007.1
NP_001159479.1
2
345
NM_004437.3
NP_004428.1
2
345
NM_203342.2
NP_976217.1
2
362
NM_203343.2
NP_976218.1
2
362
XM_005245753.1
XP_005245810.1
2
437
XM_005245757.1
XP_005245814.1
2
437
XM_005245760.1
XP_005245817.1
2
437
XM_005245761.1
XP_005245818.1
2
437
XM_005245763.1
XP_005245820.1
2
437
XM_005245764.1
XP_005245821.1
2
437
XM_005245765.1
XP_005245822.1
2
437
XM_005245768.1
XP_005245825.1
2
437
XM_005245769.1
XP_005245826.1
2
437
XM_005245770.1
XP_005245827.1
2
437
XM_005245772.4
XP_005245829.1
2
437
XM_005245773.4
XP_005245830.1
2
437
XM_005245774.1
XP_005245831.1
2
437
XM_006710434.1
XP_006710497.1
2
437
XM_006710439.1
XP_006710502.1
2
437
XM_011540956.1
XP_011539258.1
2
437
XM_011540957.1
XP_011539259.1
2
437
XM_011540958.1
XP_011539260.1
2
437
XM_011540959.1
XP_011539261.1
2
437
XM_011540960.1
XP_011539262.1
2
437
XM_011540961.1
XP_011539263.1
2
437
XM_011540962.1
XP_011539264.1
2
437
XM_011540963.2
XP_011539265.1
2
437
XM_011540964.1
XP_011539266.1
2
333
XM_011540965.2
XP_011539267.1
2
437
XM_017000581.1
XP_016856070.1
2
437
XM_017000582.1
XP_016856071.1
2
437
XM_017000583.1
XP_016856072.1
2
437
XM_017000584.1
XP_016856073.1
2
728
XM_017000585.1
XP_016856074.1
2
725
XM_017000586.1
XP_016856075.1
2
437
XM_017000587.1
XP_016856076.1
2
437
XM_017000588.1
XP_016856077.1
2
437
XM_017000589.1
XP_016856078.1
2
437
XM_017000590.1
XP_016856079.1
2
437
XM_017000591.1
XP_016856080.1
2
437
XM_017000592.1
XP_016856081.1
2
437
XM_017000593.1
XP_016856082.1
2
333
XM_017000594.1
XP_016856083.1
2
333
XM_017000595.1
XP_016856084.1
2
333
XM_017000596.1
XP_016856085.1
2
333
XM_017000597.1
XP_016856086.1
2
333
XM_017000598.1
XP_016856087.1
2
333
XM_017000599.1
XP_016856088.1
2
333
XM_017000600.1
XP_016856089.1
2
333
XM_017000601.1
XP_016856090.1
2
333
XM_017000602.1
XP_016856091.1
2
333
XM_017000603.1
XP_016856092.1
2
333
XM_017000604.1
XP_016856093.1
2
333
GenBank mRNA
AB209649.1
-
2
345
AK310655.1
-
2
378
BC039079.1
-
2
432
5167140
BM465039.1
-
2
320
DA480006.1
-
2
437
J03796.1
-
2
281
M14993.1
-
2
326
M61733.1
-
2
343
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
blood circulation
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural constituent of cytoskeleton
protein binding
calmodulin binding
1-phosphatidylinositol binding
spectrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline