This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other BYSL SIRNA Assays ›
Gene Symbol:
BYSL
Gene Name:
bystin like
Gene Aliases:
BYSTIN
Chromosome Location:
Chr.6: 41921227 - 41933046 on Build GRCh38
UniGene ID:
Hs.106880
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 147294

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: BYSL
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004053.3 NP_004044.3 4 986
GenBank mRNA AK095253.1 - 1406
AK314147.1 - 4 741
BC050645.1 - 4 986 6011459
BC062627.1 - 4 957 5226210
L36720.2 - 3 266

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

maturation of SSU-rRNA from tricistronic rRNA transcript (SSU-rRNA, 5.8S rRNA, LSU-rRNA)
trophectodermal cell differentiation
rRNA processing
cell adhesion
brain development
female pregnancy
cell proliferation
neuron differentiation
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to growth factor stimulus
cellular response to organic cyclic compound
protein binding
poly(A) RNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-pcbtx:80/100.66.76.251:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0