siRNA Details
| Gene Symbol: FNDC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001171941.2 | NP_001165412.1 | 5 | 559 | ||
| NM_153756.2 | NP_715637.2 | 5 | 625 | |||
| GenBank mRNA | AK092102.1 | - | 5 | 558 | ||
| BC062297.1 | - | 5 | 548 | 6338770 | ||
| DA393478.1 | - | 3 | 229 | |||

FNDC5
Genomic Map