This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GSE1 SIRNA Assays ›
Gene Symbol:
GSE1
Gene Name:
Gse1 coiled-coil protein
Gene Aliases:
CRHSP24, KIAA0182
Chromosome Location:
Chr.16: 85169512 - 85676206 on Build GRCh38
UniGene ID:
Hs.461647
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 148444

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: GSE1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134473.2 NP_001127945.1 2 265
NM_001278184.1 NP_001265113.1 2 319
NM_014615.3 NP_055430.1 3 538
XM_005255859.4 XP_005255916.3 4 2680
XM_005255860.3 XP_005255917.3 4 2687
XM_005255861.4 XP_005255918.3 3 2461
XM_005255863.4 XP_005255920.1 3 430
XM_005255864.4 XP_005255921.1 3 1162
XM_005255865.4 XP_005255922.1 2 943
XM_011522965.2 XP_011521267.1 3 929
XM_011522966.2 XP_011521268.1 3 396
XM_017023083.1 XP_016878572.1 3 688
GenBank mRNA BC037556.1 - 2 288 4994198
BQ214916.1 - 456
D80004.2 - 2 203
HY060424.1 - 3 541

Back To Top

More Information


Gene Ontology Categories:

Process(es)

protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline