This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other NEU4 SIRNA Assays ›
Gene Symbol:
NEU4 GAL3ST2
Gene Name:
neuraminidase 4
galactose-3-O-sulfotransferase 2
Gene Aliases:
-
GAL3ST-2, GP3ST
Chromosome Location:
Chr.2: 241809065 - 241817413 on Build GRCh38
Chromosome Location:
Chr.2: 241776825 - 241804287 on Build GRCh38
UniGene ID:
Hs.551747
Hs.632556
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID 148821

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: NEU4
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167600.2 NP_001161072.1 1 144
NM_001167601.2 NP_001161073.1 1 144
GenBank mRNA AK025617.1 - 1 16
AK096992.1 - 1 16
AK289421.1 - 1 21
AK298403.1 - 1 16
AK300969.1 - 1 17
BQ651855.1 - 1 144
DC412650.1 - 1 384
Gene Symbol: GAL3ST2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB040610.1 - 5 1484

Back To Top

More Information


Panther Classification:

Molecular Function -

hydrolase transferase

Gene Ontology Categories:

Function(s) Process(es)

glycoprotein catabolic process
glycosphingolipid metabolic process
ganglioside catabolic process
oligosaccharide catabolic process
exo-alpha-sialidase activity
protein binding
exo-alpha-(2->3)-sialidase activity
exo-alpha-(2->6)-sialidase activity
exo-alpha-(2->8)-sialidase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-kbrl8:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline