siRNA Details
| Gene Symbol: ZNF607 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172677.1 | NP_001166148.1 | 5 | 866 | ||
| NM_032689.4 | NP_116078.4 | 5 | 1217 | |||
| XM_006723435.3 | XP_006723498.1 | 5 | 1471 | |||
| XM_006723436.2 | XP_006723499.1 | 5 | 1181 | |||
| XM_006723437.2 | XP_006723500.1 | 4 | 1085 | |||
| XM_006723440.1 | XP_006723503.1 | 2 | 908 | |||
| XM_011527414.2 | XP_011525716.1 | 2 | 1162 | |||
| GenBank mRNA | AK127464.1 | - | 1 | 133 | ||
| AK131557.1 | - | 5 | 1217 | |||
| AK131559.1 | - | 5 | 866 | |||
| BC014850.1 | - | 1 | 43 | 3447451 | ||

ZNF607
Genomic Map