This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other C2ORF73 SIRNA Assays ›
Gene Symbol:
C2orf73
Gene Name:
chromosome 2 open reading frame 73
Gene Aliases:
-
Chromosome Location:
Chr.2: 54329961 - 54383673 on Build GRCh38
UniGene ID:
Hs.732626
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 149266

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: C2orf73
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100396.1 NP_001093866.1 5 749
XM_005264133.1 XP_005264190.1 4 541
XM_005264134.3 XP_005264191.1 4 624
XM_011532510.2 XP_011530812.1 5 819
XM_011532513.2 XP_011530815.1 4 722
XM_017003323.1 XP_016858812.1 5 644
XM_017003324.1 XP_016858813.1 4 707
GenBank mRNA AK097617.1 - 4 541
BC031669.1 - 5 712
BC126244.1 - 4 510 8991960
BC126246.1 - 4 510 8991962
BC144191.1 - 3 394
BC144192.1 - 5 544 9052712

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline