This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other FAM230B SIRNA Assays ›
Gene Symbol:
FAM230B LOC100996335 LOC100996432 LOC105369189
Gene Name:
family with sequence similarity 230 member B (non-protein coding)
uncharacterized LOC100996335
uncharacterized LOC100996432
uncharacterized LOC105369189
Gene Aliases:
-
-
-
-
Chromosome Location:
Chr.22: 21166903 - 21192156 on Build GRCh38
Chromosome Location:
Chr.22: 21301260 - 21326636 on Build GRCh38
Chromosome Location:
Chr.22: 18865043 - 18894431 on Build GRCh38
Chromosome Location:
Chr.22|NT_187386: 110221 - 127881 on Build GRCh38
UniGene ID:
-
Hs.645542
Hs.631668
-
Species:
Human
Species Specific ID (Flybase ID):
-
-
-
-
siRNA ID 149394

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: FAM230B
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_108107.1 9 1397
GenBank mRNA AK128837.1 - 3 1210
Gene Symbol: LOC100996335
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136559.1 8 1182
Gene Symbol: LOC100996432
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_136571.1 8 1262
Gene Symbol: LOC105369189
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017030128.1 XP_016885617.1 8 2058

Back To Top

More Information



Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0