This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other TEX19 SIRNA Assays ›
Gene Symbol:
TEX19
Gene Name:
testis expressed 19
Gene Aliases:
-
Chromosome Location:
Chr.17: 82359247 - 82363776 on Build GRCh38
UniGene ID:
Hs.231897
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 149927

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: TEX19
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207459.3 NP_997342.1 2 1695
XM_017024638.1 XP_016880127.1 2 1754
XM_017024639.1 XP_016880128.1 2 1782
GenBank mRNA AK093086.1 - 2 1698
BC016939.2 - 1 1656 4419895
BC036191.1 - 2 1695 5297531
BC050391.1 - 2 1672 5745215
BC057820.1 - 2 1696 4837469

Back To Top

More Information


Gene Ontology Categories:

Function(s)

spermatogenesis
cell differentiation
meiotic cell cycle

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-hssmq:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline