This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SKIL SIRNA Assays ›
Gene Symbol:
Skil
Gene Name:
SKI-like
Gene Aliases:
Skir, SnoN, sno
Chromosome Location:
Chr.3: 31095058 - 31122923 on Build GRCm38
UniGene ID:
Mm.15406
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 151701

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Skil
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039090.2 NP_001034179.1 2 937
NM_011386.3 NP_035516.2 2 937
GenBank mRNA AB221599.1 - 1 214
AK087797.1 - 2 886
AK154471.1 - 2 884
AK162952.1 - 2 936
AK163526.1 - 2 864
AK171402.1 - 2 886
BC049934.1 - 1 363 5387475
U10530.1 - 1 708
U10532.1 - 1 708
U14655.1 - 1 270

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
blastocyst formation
lymphocyte homeostasis
transcription from RNA polymerase II promoter
cell cycle arrest
transforming growth factor beta receptor signaling pathway
skeletal muscle tissue development
negative regulation of transforming growth factor beta receptor signaling pathway
negative regulation of cell differentiation
positive regulation of axonogenesis
protein homotrimerization
protein heterotrimerization
lens fiber cell differentiation
response to growth factor
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of intrinsic apoptotic signaling pathway in response to DNA damage
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
transcription corepressor activity
protein binding
protein domain specific binding
protein complex binding
SMAD binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-bwr29:80/100.66.78.44:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0