siRNA Details
| Gene Symbol: Slfn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF099975.1 | - | 1 | 409 | ||
| AF099976.1 | - | 2 | 1501 | |||
| AK137559.1 | - | 8 | 3147 | |||
| AK137689.1 | - | 7 | 3068 | |||
| BC044865.1 | - | 8 | 3155 | 4459564 | ||
| BC046511.1 | - | 3 | 1034 | |||
| BC150770.1 | - | 5 | 2762 | 9087681 | ||

Slfn4
Genomic Map