This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other STAR SIRNA Assays ›
Gene Symbol:
Star
Gene Name:
steroidogenic acute regulatory protein
Gene Aliases:
AV363654, D8Ertd419e
Chromosome Location:
Chr.8: 25808513 - 25815982 on Build GRCm38
UniGene ID:
Mm.293314
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 151870

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Star
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011485.4 NP_035615.2 4 415
GenBank mRNA AK014431.1 - 4 423
AK019725.2 - 4 455
AK054470.1 - 4 415
AK136327.1 - 4 425
BC082283.1 - 4 426 30436512
L36062.1 - 4 431

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
bile acid biosynthetic process
estrogen biosynthetic process
transport
lipid transport
glucocorticoid metabolic process
positive regulation of gene expression
cellular response to interferon-alpha
negative regulation of neuron apoptotic process
cellular lipid metabolic process
regulation of neuronal synaptic plasticity
positive regulation of neurogenesis
regulation of steroid biosynthetic process
lipid binding
cholesterol binding
cholesterol transporter activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0