siRNA Details
| Gene Symbol: Optc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001160420.2 | NP_001153892.1 | 7 | 978 | ||
| NM_001160421.2 | NP_001153893.1 | 6 | 1089 | |||
| NM_054076.3 | NP_473417.2 | 7 | 1248 | |||
| GenBank mRNA | AF333980.1 | - | 7 | 1247 | ||
| AK153750.1 | - | 7 | 979 | |||
| AY077682.1 | - | 7 | 1240 | |||
| BC014874.1 | - | 2 | 941 | |||
| BC115928.1 | - | 6 | 1042 | 40104738 | ||

Optc
Genomic Map