siRNA Details
| Gene Symbol: Il17d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145837.3 | NP_665836.2 | 3 | 1066 | ||
| GenBank mRNA | AF458063.1 | - | 2 | 988 | ||
| AK131917.1 | - | 3 | 1115 | |||
| BC099514.1 | - | 1 | 508 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Il17d| Gene Symbol: Il17d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145837.3 | NP_665836.2 | 3 | 1066 | ||
| GenBank mRNA | AF458063.1 | - | 2 | 988 | ||
| AK131917.1 | - | 3 | 1115 | |||
| BC099514.1 | - | 1 | 508 | |||