This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other E330021D16RIK SIRNA Assays ›
Gene Symbol:
E330021D16Rik
Gene Name:
RIKEN cDNA E330021D16 gene
Gene Aliases:
C85549
Chromosome Location:
Chr.6: 136400312 - 136419868 on Build GRCm38
UniGene ID:
Mm.21182
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 153844

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: E330021D16Rik
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001201390.1 NP_001188319.1 2 1033
GenBank mRNA AK054382.1 - 2 1034
AK135843.1 - 2 1034
BC099944.1 - 2 1024

Back To Top

More Information


Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-vc5lg:80/100.66.78.61:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0