This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EPB4.1 SIRNA Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte protein band 4.1
Gene Aliases:
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056
Chromosome Location:
Chr.4: 131923413 - 132075579 on Build GRCm38
UniGene ID:
Mm.30038
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 154827

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Epb4.1
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128606.1 NP_001122078.1 2 385
NM_001128607.1 NP_001122079.1 2 295
NM_183428.3 NP_906273.3 2 1647
GenBank mRNA AB370261.1 - 1 170
AK220462.1 - 2 292
BC068138.1 - 2 255
BC079875.1 - 2 214 6844082
CA324069.1 - 2 394
CB723734.1 - 2 214
CJ109236.1 - 2 295
L00919.1 - 2 1641

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

regulation of cell shape
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
calmodulin binding
1-phosphatidylinositol binding
cytoskeletal protein binding
spectrin binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-f8k5r:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline