siRNA Details
| Gene Symbol: Mrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008626.3 | NP_032652.3 | 8 | 1556 | ||
| GenBank mRNA | AK028157.1 | - | 8 | 1547 | ||
| BC116642.1 | - | 8 | 1482 | 40086489 | ||
| U56734.1 | - | 8 | 1519 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mrc2| Gene Symbol: Mrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008626.3 | NP_032652.3 | 8 | 1556 | ||
| GenBank mRNA | AK028157.1 | - | 8 | 1547 | ||
| BC116642.1 | - | 8 | 1482 | 40086489 | ||
| U56734.1 | - | 8 | 1519 | |||