siRNA Details
| Gene Symbol: Smc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301412.1 | NP_001288341.1 | 21 | 3104 | ||
| NM_008017.4 | NP_032043.3 | 21 | 3028 | |||
| GenBank mRNA | AJ534939.1 | - | 20 | 2859 | ||
| AK145341.1 | - | 21 | 3058 | |||
| BC071232.1 | - | 6 | 763 | |||
| BC094380.1 | - | 21 | 3000 | 30543190 | ||
| U42385.1 | - | 4 | 471 | |||

Smc2
Genomic Map