This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other GDF3 SIRNA Assays ›
Gene Symbol:
Gdf3
Gene Name:
growth differentiation factor 3
Gene Aliases:
C78318, Gdf-3, Vgr-2, Vgr2, ecat9
Chromosome Location:
Chr.6: 122605403 - 122610071 on Build GRCm38
UniGene ID:
Mm.299742
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 157741

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Gdf3
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008108.4 NP_032134.2 1 252
GenBank mRNA AK082877.1 - 1 172
AK146174.1 - 1 178
AK152457.1 - 1 253
AK160533.1 - 1 210
AK167962.1 - 1 178
BC101963.1 - 1 167 40045099
BC101964.1 - 1 167 40045100
BC101965.1 - 1 167
BC103565.1 - 1 166 40045101
L06443.1 - 1 268
S52658.1 - 1 208

Back To Top

More Information


Panther Classification:

Molecular Function -

growth factor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
eye development
in utero embryonic development
response to dietary excess
signal transduction
endoderm development
mesoderm development
regulation of cell fate commitment
positive regulation of pathway-restricted SMAD protein phosphorylation
BMP signaling pathway
negative regulation of BMP signaling pathway
notochord development
somite rostral/caudal axis specification
growth
regulation of apoptotic process
regulation of MAPK cascade
negative regulation of epidermal cell differentiation
negative regulation of myoblast differentiation
cell development
formation of anatomical boundary
SMAD protein signal transduction
primitive streak formation
cytokine activity
transforming growth factor beta receptor binding
protein binding
growth factor activity
protein kinase binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gfvrm:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline