This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other H3F3A SIRNA Assays ›
Gene Symbol:
H3f3a Gm12657
Gene Name:
H3 histone, family 3A
predicted gene 12657
Gene Aliases:
H3.3A
OTTMUSG00000007855
Chromosome Location:
Chr.1: 180802568 - 180813603 on Build GRCm38
Chromosome Location:
Chr.4: 94593650 - 94600319 on Build GRCm38
UniGene ID:
Mm.138832
Mm.449778
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
siRNA ID 157970

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: H3f3a
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008210.4 NP_032236.1 4 567
GenBank mRNA AK008405.1 - 3 468
AK037900.1 - 4 569
AK150467.1 - 4 527
AK150468.1 - 4 527
AK150591.1 - 4 522
AK150928.1 - 4 522
AK151634.1 - 4 527
AK152293.1 - 4 522
AK153034.1 - 4 526
AK160677.1 - 4 526
AK162040.1 - 4 525
AK167879.1 - 4 526
AK168197.1 - 4 526
AK169608.1 - 4 522
AK187043.1 - 1 63
BC002268.1 - 4 512 3495830
BC012687.1 - 4 516 4224280
BC083353.1 - 4 507 5714859
BC088835.1 - 4 504 6437687
BC106177.1 - 4 501 6511787
Z85979.1 - 4 515
Gene Symbol: Gm12657
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081019.1 NP_001074488.1 2 454
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BC027275.1 - 2042 Chr. - - [-] on Build

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_003750348, XM_002729644, XM_002728043, FQ218544, FQ220271, and others.

Panther Classification:

Molecular Function -

histone

Gene Ontology Categories:

Function(s) Process(es)

osteoblast differentiation
muscle cell differentiation
negative regulation of chromosome condensation
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
nucleosomal DNA binding
histone binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-c565d5cb7-nqkv9:80/100.66.76.40:80.
git-commit: 04d74f786bdbf98a9bc1b2a22061fbbfd0e8b2c2
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.38.0-2025.10.43-1.0