siRNA Details
| Gene Symbol: Idh3g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008323.1 | NP_032349.1 | 8 | 677 | ||
| GenBank mRNA | AJ748655.1 | - | 8 | 629 | ||
| AK156309.1 | - | 8 | 675 | |||
| AK166925.1 | - | 7 | 634 | |||
| AK167131.1 | - | 5 | 1395 | |||
| AK168526.1 | - | 8 | 675 | |||
| BC085179.1 | - | 8 | 650 | 30699000 | ||
| BC119170.1 | - | 8 | 660 | 8733919 | ||
| BC119172.1 | - | 8 | 660 | 8733921 | ||
| U68564.1 | - | 8 | 677 | |||

Idh3g
Genomic Map