This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other IGFALS SIRNA Assays ›
Gene Symbol:
Igfals
Gene Name:
insulin-like growth factor binding protein, acid labile subunit
Gene Aliases:
ALS, Albs, mKIAA4111
Chromosome Location:
Chr.17: 24877653 - 24882011 on Build GRCm38
UniGene ID:
Mm.3135
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158249

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Igfals
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008340.3 NP_032366.2 2 2012
GenBank mRNA AK004926.1 - 2 2014
AK220290.1 - 2 2524
BC129875.1 - 2 2001 40110389
BC129876.1 - 2 2001 40110391

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
protein binding
insulin-like growth factor binding
protein C-terminus binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-mjsjv:80/100.66.76.16:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline