This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other EPHA2 SIRNA Assays ›
Gene Symbol:
Epha2
Gene Name:
Eph receptor A2
Gene Aliases:
AW545284, Eck, Myk2, Sek-2, Sek2
Chromosome Location:
Chr.4: 141301221 - 141329384 on Build GRCm38
UniGene ID:
Mm.2581
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158404

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Epha2
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010139.3 NP_034269.2 3 317
GenBank mRNA AK137704.1 - 3 299
AK144202.1 - 3 301
BC140960.1 - 3 273 9055572
BY024029.1 - 3 298
CJ088701.1 - 3 321
U07634.1 - 3 281
X78339.1 - 3 279

Back To Top

More Information


Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
angiogenesis
blood vessel development
vasculogenesis
osteoblast differentiation
protein phosphorylation
apoptotic process
cell adhesion
transmembrane receptor protein tyrosine kinase signaling pathway
intrinsic apoptotic signaling pathway in response to DNA damage
regulation of lamellipodium assembly
notochord formation
phosphorylation
cell migration
peptidyl-tyrosine phosphorylation
neural tube development
cell differentiation
neuron differentiation
keratinocyte differentiation
osteoclast differentiation
mammary gland epithelial cell proliferation
regulation of cell adhesion mediated by integrin
post-anal tail morphogenesis
protein kinase B signaling
regulation of blood vessel endothelial cell migration
regulation of angiogenesis
bone remodeling
ephrin receptor signaling pathway
axial mesoderm formation
notochord morphogenesis
negative regulation of protein kinase B signaling
notochord cell development
cell chemotaxis
branching involved in mammary gland duct morphogenesis
lens fiber cell morphogenesis
regulation of ERK1 and ERK2 cascade
response to growth factor
positive regulation of establishment of protein localization to plasma membrane
activation of GTPase activity
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
ephrin receptor activity
protein binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0