This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other PDPN SIRNA Assays ›
Gene Symbol:
Pdpn
Gene Name:
podoplanin
Gene Aliases:
Gp38, OTS-8, RANDAM-2, T1-alpha, T1a, T1alpha
Chromosome Location:
Chr.4: 143267409 - 143299564 on Build GRCm38
UniGene ID:
Mm.2976
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 158798

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Pdpn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010329.3 NP_034459.2 4 697
GenBank mRNA AJ250246.1 - 4 426
AJ297944.2 - 4 403
AK158855.1 - 4 697
BC026551.1 - 4 581 4985288
M73748.1 - 4 554
M96645.1 - 4 464

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
lymphangiogenesis
prostaglandin metabolic process
water transport
amino acid transport
movement of cell or subcellular component
cell adhesion
signal transduction
multicellular organism development
cell proliferation
regulation of cell shape
positive regulation of platelet activation
folic acid transport
single organismal cell-cell adhesion
lung development
positive regulation of cell migration
tube morphogenesis
lung alveolus development
positive regulation of cellular component movement
visceral serous pericardium development
regulation of G1/S transition of mitotic cell cycle
water transmembrane transporter activity
folic acid transporter activity
amino acid transmembrane transporter activity
water channel activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline