siRNA Details
| Gene Symbol: Hoxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010449.4 | NP_034579.3 | 1 | 337 | ||
| GenBank mRNA | AK083575.1 | - | 1 | 338 | ||
| AK143943.1 | - | 1 | 332 | |||
| BC138097.1 | - | 1 | 271 | 8861116 | ||
| BC138098.1 | - | 1 | 274 | 8861117 | ||
| M22115.1 | - | 1 | 306 | |||

Hoxa1
Genomic Map