This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HOXD9 SIRNA Assays ›
Gene Symbol:
Hoxd9
Gene Name:
homeobox D9
Gene Aliases:
Hox-4.4, Hox-5.2
Chromosome Location:
Chr.2: 74697763 - 74700208 on Build GRCm38
UniGene ID:
Mm.26544
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 159297

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hoxd9
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013555.4 NP_038583.1 2 1735
GenBank mRNA AK134427.1 - 2 1525
AK144746.1 - 2 880
BC019150.1 - 2 1770 5064578

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
multicellular organism development
single fertilization
skeletal muscle tissue development
adult locomotory behavior
anterior/posterior pattern specification
proximal/distal pattern formation
regulation of gene expression
mammary gland development
embryonic forelimb morphogenesis
forelimb morphogenesis
hindlimb morphogenesis
positive regulation of transcription from RNA polymerase II promoter
embryonic skeletal system morphogenesis
embryonic skeletal system development
peripheral nervous system neuron development
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-7669c5b596-lsgr5:80/100.66.76.41:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0