This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other HPRT SIRNA Assays ›
Gene Symbol:
Hprt
Gene Name:
hypoxanthine guanine phosphoribosyl transferase
Gene Aliases:
C81579, HPGRT, Hprt1
Chromosome Location:
Chr.X: 52988078 - 53021660 on Build GRCm38
UniGene ID:
Mm.299381
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 159299

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 283.00
Your Price
Online offer: 209.42
209.42
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Hprt
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013556.2 NP_038584.2 3, 4 465
GenBank mRNA AK002286.1 - 3, 4 410
AK082827.1 - 3, 4 355
AK088114.1 - 3, 4 355
AK146626.1 - 3, 4 411
BC004686.1 - 3, 4 380 3500885
BC083145.1 - 3, 4 351 30445781
J00423.1 - 3, 4 406

Back To Top

More Information


Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

response to amphetamine
purine nucleotide biosynthetic process
purine ribonucleoside salvage
adenine salvage
guanine salvage
grooming behavior
locomotory behavior
nucleoside metabolic process
cytolysis
striatum development
cerebral cortex neuron differentiation
central nervous system neuron development
GMP salvage
IMP salvage
dopamine metabolic process
hypoxanthine salvage
positive regulation of dopamine metabolic process
GMP catabolic process
IMP metabolic process
adenine metabolic process
hypoxanthine metabolic process
lymphocyte proliferation
dendrite morphogenesis
protein homotetramerization
nucleotide binding
magnesium ion binding
hypoxanthine phosphoribosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
identical protein binding
protein homodimerization activity
metal ion binding
guanine phosphoribosyltransferase activity

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-fbxsq:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline