This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other AMBN SIRNA Assays ›
Gene Symbol:
Ambn
Gene Name:
ameloblastin
Gene Aliases:
-
Chromosome Location:
Chr.5: 88455991 - 88468531 on Build GRCm38
UniGene ID:
Mm.8437
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID 160652

Purification
Size
Availability Inventoried
Catalog # AM16708
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: Ambn
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK017250.1 - 3, 4 252
BC087927.1 - 3, 4 252 30504437
U65021.1 - 3, 4 232

Back To Top

More Information


Other Species Targeted

This siRNA also targets the following transcripts in Human: NM_016519, BC106931, BC106932, AJ537437, AK314918, and others.
This siRNA also targets the following transcripts in Rat: NM_012900, U35097, Z50083.

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-dz54l:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline