siRNA Details
| Gene Symbol: Anp32a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009672.3 | NP_033802.2 | 2, 3 | 370 | ||
| GenBank mRNA | AF022957.1 | - | 2, 3 | 279 | ||
| AK020753.1 | - | 2, 3 | 332 | |||
| BC062899.1 | - | 2, 3 | 285 | 6415168 | ||
| U73478.1 | - | 2, 3 | 237 | |||
| Gene Symbol: Anp32-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AY702092.1 | - | 1 | 212 | ||

Anp32a
Genomic Map