siRNA Details
| Gene Symbol: HIST1H1T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005323.3 | NP_005314.2 | 1 | 333 | ||
| GenBank mRNA | BC069517.1 | - | 1 | 333 | 7262183 | |
| BC130350.1 | - | 1 | 326 | 40146381 | ||
UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.

| Gene Symbol: HIST1H1T | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_005323.3 | NP_005314.2 | 1 | 333 | ||
| GenBank mRNA | BC069517.1 | - | 1 | 333 | 7262183 | |
| BC130350.1 | - | 1 | 326 | 40146381 | ||
HIST1H1T
Genomic Map