siRNA Details
| Gene Symbol: Chrd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278041.1 | NP_001264970.1 | 11 | 1366 | ||
| NM_009893.2 | NP_034023.1 | 11 | 1300 | |||
| GenBank mRNA | AF069501.1 | - | 11 | 1300 | ||
| AF096276.1 | - | 11 | 1279 | |||
| AK077460.1 | - | 12 | 1402 | |||
| AK149445.1 | - | 11 | 1275 | |||
| AK165339.1 | - | 11 | 1293 | |||
| BC145017.1 | - | 10 | 1507 | |||
| BC145018.1 | - | 12 | 1428 | |||
| BC145019.1 | - | 12 | 1406 | |||
| BC145696.1 | - | 11 | 1364 | 40130999 | ||

Chrd
Genomic Map