siRNA Details
| Gene Symbol: Aebp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178803.2 | NP_848918.1 | 4 | 870 | ||
| GenBank mRNA | AB221647.1 | - | 4 | 521 | ||
| AF090326.1 | - | 605 | ||||
| AK029011.1 | - | 4 | 872 | |||
| AK045838.1 | - | 4 | 636 | |||
| AK045987.1 | - | 4 | 636 | |||
| AK155272.1 | - | 4 | 602 | |||
| BC025467.1 | - | 4 | 1328 | 5351959 | ||
| BC031376.1 | - | 5 | 731 | 5356616 | ||
| BC057681.1 | - | 4 | 650 | 3499299 | ||

Aebp2
Genomic Map