This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
See other SNCAIP SIRNA Assays ›
Gene Symbol:
SNCAIP
Gene Name:
synuclein alpha interacting protein
Gene Aliases:
SYPH1, Sph1
Chromosome Location:
Chr.5: 122311354 - 122464219 on Build GRCh38
UniGene ID:
Hs.426463
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID 16263

Purification
Size
Availability Inventoried
Catalog # AM16708
Price 4,126.00
Your Price
Online offer:
4126.0
Check your price ›
  • Genomic Map
  • siRNA Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

siRNA Details

Gene Symbol: SNCAIP
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308100.1 NP_001295029.1 6 1908
NM_001308105.1 NP_001295034.1 3 247
NM_005460.3 NP_005451.2 4 463
NR_131761.1 4 388
XM_005272138.3 XP_005272195.1 4 386
XM_005272139.1 XP_005272196.1 4 326
XM_006714734.2 XP_006714797.1 4 519
XM_011543737.2 XP_011542039.1 5 521
XM_011543738.2 XP_011542040.1 5 520
XM_011543739.1 XP_011542041.1 5 446
XM_011543741.2 XP_011542043.1 5 470
XM_011543743.2 XP_011542045.1 5 895
XM_011543745.1 XP_011542047.1 4 350
XM_017010078.1 XP_016865567.1 5 2848
XM_017010079.1 XP_016865568.1 5 521
XM_017010080.1 XP_016865569.1 5 521
XM_017010081.1 XP_016865570.1 5 519
XM_017010082.1 XP_016865571.1 4 350
XM_017010084.1 XP_016865573.1 5 527
XM_017010086.1 XP_016865575.1 5 525
GenBank mRNA AB110788.1 - 3 201
AB110789.1 - 3 201
AB110790.1 - 4 342
AF076929.1 - 4 294
AK310835.1 - 4 295
BC040552.1 - 6 1770 5298588

Back To Top

More Information


Gene Ontology Categories:

Function(s) Process(es)

cell death
dopamine metabolic process
cellular protein metabolic process
regulation of neurotransmitter secretion
regulation of inclusion body assembly
protein binding
ubiquitin protein ligase binding
identical protein binding

Back To Top

Related Products

  • Anti-GAPDH, Mouse Monoclonal 6C5
  • Anti-β-Actin, Mouse Monoclonal AC-15
  • TaqMan® Gene Expression Cells-to-CT™
  • Neon™ Transfection System 100 µL Kit
  • Lipofectamine™ RNAiMAX Transfection Reagent
  • Invivofectamine 2.0
  • Lipofectamine™ 2000 Transfection Reagent
  • Neon™ Transfection System

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0